missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa, APC, clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 17-4714-42

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12642059

  • £93.49 / Each


EU Stock 33
Add to basket

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control.Applications Reported: APC Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis.Applications Tested: The Mouse IgG1 K Isotype Control is offered in 2 formats:- μg size: This can be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by intracellular staining for flow cytometric analysis. This can be used at test size: 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1, K
Control
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
P3.6.2.8.1
APC
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.