Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 53-4714-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: Alexa Fluor™ 488 Mouse IgG1, K Isotype Control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: This Mouse IgG1 K Isotype Control is offered in 2 formats: - μg size: has been tested. This can be used at at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested. This can be used at test size: 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
Alexa Fluor 488 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.