Learn More
IgG1/IgG2a Mouse Anti-Rat, Brilliant Violet 510, Clone: G28-5, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743850
Additional Details : Weight : 0.01000kg
Descripción
The G28-5 monoclonal antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. A suspension of pooled rat IgG1 was used as the source of immunogen.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.Especificaciones
IgG1/IgG2a | |
G28-5 | |
Brilliant Violet 510 | |
IgG1/IgG2a, Ig gamma-1/2a, Immunoglobulin gamma 1/2a | |
Pooled Rat IgG | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a |