missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
£76.50 - £178.00
Specifications
Antigen | Armenian Hamster IgG |
---|---|
Clone | eBio299Arm |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15341330
|
Affymetrix eBioscience
46-4888-80 |
25 μg |
£76.50
25µg |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
15351330
|
Affymetrix eBioscience
46-4888-82 |
100 μg |
£178.00
100µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. Use isotype control at the same concentration as experimental antibody. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Armenian Hamster IgG Control antibody specifically detects Armenian Hamster IgG in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Armenian Hamster IgG | |
0.2 mg/mL | |
PerCP-eFluor 710 | |
Armenian Hamster | |
Not Applicable | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBio299Arm | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG | |
Affinity chromatography |
For Research Use Only.