Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), PE-eFluor™ 610, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
£190.00
Specifications
Antigen | Armenian Hamster IgG |
---|---|
Clone | eBio299Arm |
Conjugate | PE-eFluor 610 |
Form | Liquid |
Host Species | Armenian Hamster |
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes.Use the isotype control at the same concentration as the experimental antibody. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Armenian Hamster IgG Control antibody specifically detects Armenian Hamster IgG in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Armenian Hamster IgG | |
PE-eFluor 610 | |
Armenian Hamster | |
PBS with 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eBio299Arm | |
Liquid | |
RUO | |
IgG | |
Affinity chromatography |
For Research Use Only.