missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, PE-Cyanine5, clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Isotype Control

Brand:  Affymetrix eBioscience 15-4888-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15360520

  • £74.49 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis. Isotype control should be used at the same concentration as experimental antibody.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG
eBio299Arm
PE-Cyanine5
Conjugated
IgG
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Armenian Hamster
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Videos