missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, eFluor 660, clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Isotype Control

Brand:  Affymetrix eBioscience 50-4888-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15559966

  • £43.63 / 25Āµg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This eBio299Arm antibody has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis.Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use isotype control at the same concentration as experimental antibody.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG
IgG
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow CytometryĆ¾ImmunocytochemistryĆ¾Immunohistochemistry
eBio299Arm
eFluor 660
Conjugated
Armenian Hamster
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.