Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 506, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
£55.75 - £150.00
Specifications
Antigen | Armenian Hamster IgG |
---|---|
Clone | eBio299Arm |
Conjugate | eFluor 506 |
Form | Liquid |
Host Species | Armenian Hamster |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15540687
|
Affymetrix eBioscience
69-4888-82 |
100 μg |
£150.00
100µg |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15530687
|
Affymetrix eBioscience
69-4888-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Armenian Hamster IgG Control antibody specifically detects Armenian Hamster IgG in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Armenian Hamster IgG | |
eFluor 506 | |
Armenian Hamster | |
IgG | |
Affinity chromatography |
eBio299Arm | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.