missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-6 Rat anti-Mouse, Clone: 122312, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | IGFBP-6 |
---|---|
Clone | 122312 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6 Gly24-Gly238 Accession # P47880 |
Gene ID (Entrez) | 3489 |
Gene Accession No. | P47880 |
Description
IGFBP-6 Monoclonal specifically detects IGFBP-6 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGFBP-6 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6 Gly24-Gly238 Accession # P47880 | |
P47880 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
122312 | |
3489 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse IGFBP-6 in ELISAs and Western blots. In Western blots, 10% cross-reactivity with recombinant human (rh) IGFBP-3 and rhIGFBP-5 is observed and no cross-reactivity with rhIGFBP-1, recombinant mouse (rm) IGFBP-1, rhIGFBP-2, rmIGFBP-2, rmIGFBP-3, rhIGFBP-4, rmIGFBP-5, or rhIGFBP-6 is observed. | |
Unconjugated | |
Western Blot, ELISA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only