missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-1 Rat anti-Mouse, Clone: 176110, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | IGFBP-1 |
---|---|
Clone | 176110 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-1 Ala26-Asn272 Accession # P47876 |
Gene ID (Entrez) | 3484 |
Gene Accession No. | P47876 |
Description
IGFBP-1 Monoclonal specifically detects IGFBP-1 in Mouse samples. It is validated for Western Blot.Specifications
IGFBP-1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-1 Ala26-Asn272 Accession # P47876 | |
P47876 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
176110 | |
3484 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse IGFBP-1 in direct ELISAs and Western blots. In Western blots, weak cross-reactivity with recombinant mouse (rm) IGFBP-5 is observed and no cross-reactivity with recombinant human (rh) IGFBP-1, rmIGFBP-2, rmIGFBP-3, rhIGFBP-4, rmIGFBP-6, or rhIGFBP-7 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only