missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549760
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total IGF2BP3 protein.
IGF2BP3 Polyclonal antibody specifically detects IGF2BP3 in Human, Mouse samples. It is validated for Western Blot.Specifications
IGF2BP3 | |
Polyclonal | |
Unconjugated | |
IGF2BP3 | |
2610101N11Rik; AA522010; Ab2-255; AL022933; AU045931; cancer/testis antigen 98; CT98; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; insulin-like growth factor 2, binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; Koc13; mimp3; mIMP-3; Neilsen; RGD1306512; VICKZ family member 3; Vickz3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10643, 140488 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00425, Q9CPN8 | |
IGF2BP3 | |
Synthesized peptide derived from N-terminal of human IGF2BP3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |