Learn More
IgE Monoclonal Antibody (23G3), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£80.75 - £169.00
Specifications
Antigen | IgE |
---|---|
Clone | 23G3 |
Concentration | 0.5 mg/mL |
Applications | ELISA, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15269989
|
Affymetrix eBioscience
13-5992-81 |
50 μg |
£80.75
50µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15279989
|
Affymetrix eBioscience
13-5992-82 |
100 μg |
£169.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 23G3 monoclonal antibody reacts with the epsilon heavy chain of mouse IgE. It does not react with other classes of mouse immunoglobulin including IgD, IgG, IgA, or IgM. Applications Reported: The 23G3 antibody has been reported for use in flow cytometric analysis, and ELISA. Applications Tested: The 23G3 antibody has been tested by ELISA and can be used at 1:5000-1:10000. This 23G3 antibody has also been tested by flow cytometric analysis of IGEL cells, and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.Specifications
IgE | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Gm900; Ighe; Immunoglobulin E; Immunoglobulin heavy constant epsilon; ImmunoglobulinE | |
Ighe | |
Primary | |
4° C, do not freeze | |
Ighe |
23G3 | |
ELISA, Flow Cytometry | |
Biotin | |
Rat | |
Mouse | |
P06336 | |
380792 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.