missing translation for 'onlineSavingsMsg'
Learn More

IgE Mouse anti-Human, FITC, Clone: Ige21, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-6986-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15576576

  • £58.50 / 25 tests

EU Stock 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
Specifications

Specifications

IgE
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01854
IGHE
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3497
Flow Cytometry
Ige21
FITC
IGHE
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Documents

For Research Use Only.