missing translation for 'onlineSavingsMsg'
Learn More

IgD Mouse anti-Human, Super Bright 702, Clone: IA6-2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen 67-9868-41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943784

  • £125.00 / Pack of 25

Estimated Shipment
21-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predomit (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta. Applications Reported: This G14D9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G14D9 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Specifications

Specifications

IgD
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
IA6-2
Super Bright 702
IGHD
IGHD
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3495
Videos
SDS
Documents