missing translation for 'onlineSavingsMsg'
Learn More

IgD Monoclonal Antibody (11-26c (11-26)), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5993-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15508616

  • £97.50 / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

IgD
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01881
Ighd
Affinity chromatography
RUO
380797
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
11-26c (11-26)
PE-Cyanine7
Ighd
IgD; Igh-5; IGHD; immunoglobulin heavy constant delta
Rat
25 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.