missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgD, FITC, clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5993-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258329

  • £136.41 / 500µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.Applications Reported: The 11-26c antibody has been reported for use in flow cytometric analysis to detect the surface IgD on mouse B cells.Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgD
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Ighd
IgG2a, kappa
500μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
380797
Flow Cytometry
11-26c (11-26)
FITC
Ighd
Immunoglobulin D, Ig D
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.