missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgD, eFluor 450, clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5993-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15361820

  • £125.48 / 100µg


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis.Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgD
11-26c (11-26)
eFluor 450
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Immunoglobulin D, Ig D
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.