Learn More
IgD Monoclonal Antibody (11-26c (11-26)), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£82.00 - £166.00
Specifications
Antigen | IgD |
---|---|
Clone | 11-26c (11-26) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15351820
|
Affymetrix eBioscience
48-5993-80 |
25 μg |
£82.00
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15361820
|
Affymetrix eBioscience
48-5993-82 |
100 μg |
£166.00
100µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.Specifications
IgD | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgD; Igh-5; IGHD; immunoglobulin heavy constant delta | |
Ighd | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ighd |
11-26c (11-26) | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
P01881 | |
380797 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.