Learn More
Epredia™ IgA Antibody
Designed for immunofluorescence staining, Epredia™ IgA, FITC Labeled Antibody allows rapid and inexpensive assessment of tissues.
Brand: Epredia™ SH-1920-R2
Code : LD
Additional Details : Weight : 0.00400kg
Description
Host Species: Sheep
Species Reactivity: Recognizes the α-chain of human IgA. Cross-reactivity seen with bovine, mouse and rat serum.
Immunogen: IgA isolated from human serum
Positive Control: Normal tonsil fixed and processed in the same manner as the test specimen
Cellular Localization: Aggregates found in tonsil, lymph node and spleen
Recommended for:
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
IgA | |
Polyclonal | |
FITC | |
IgA isolated from human serum | |
IVD | |
Primary | |
Prediluted |
Immunofluorescence | |
Polyclonal | |
Sheep | |
2 mL | |
Cancer and Tumor Biology | |
Human, Mouse |