missing translation for 'onlineSavingsMsg'
Learn More

Lambda light chain Monoclonal Antibody (1-155-2), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9990-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15548296

  • £220.00 / 100 tests
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody 1-155-2 recognizes the lambda light chain of human immunoglobulin. Approximately 22-kDa, the Ig lambda chain is comprised of one variable and one constant region. Two lambda chains together with the heavy chain comprise the whole Ig structure. Expression of Ig lambda is typically found in less than half of B lymphocytes, and has been associated with neoplastic lymphomas. Applications Reported: This 1-155-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1-155-2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Lambda light chain Monoclonal antibody specifically detects Lambda light chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Lambda light chain
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P01701, P0CG04
IGL, IGLC1, IGLV, IGLV1-51
Affinity chromatography
RUO
28820, 3535, 3537, 3546
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
1-155-2
APC
IGL
Bence Jones Protein; BJP; Clambda1; Ig lambda chain V-I region BL2; IgL; Igl@; IGLC; IGLC 1/2/3; IGLC1; Igl-C1; IGLL5; IGLV; IGLV@; immmunoglobulin lambda light chain variable region; Immunoglobulin L; immunoglobulin lambda chain complex; immunoglobulin lambda constant 1; immunoglobulin lambda constant 7; immunoglobulin lambda light chain; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; immunoglobulin lambda variable cluster; Lambda LC; lambda-immunoglobulin; LOC100060608; Mcg Marker; Paraprotein
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.