missing translation for 'onlineSavingsMsg'
Learn More

Human Kappa Light Chain, PE, clone: TB28-2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9970-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15537296

  • £62.63 / 25 tests


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody TB28-2 recognizes the kappa light chain of human immunoglobulin. Approximately 22-kDa, the Ig kappa chain is comprised of one variable and one constant region. Two kappa chains together with the heavy chain comprise the whole Ig structure. Expression of Ig kappa is typically found on more than half of B lymphocytes.Applications Reported: This TB28-2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TB28-2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ig kappa light chain
TB28-2
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Immunoglobulin, Ig k
IgG1
RUO
Primary
Human
Videos