Learn More
IFT81 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553921
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KVVSEPAMGH SDLLELESKI NEINTEINQL IEKKMMRNEP IEGKLSLYRQ QASIISRKKE AKAEELQEAK EKLASLEREA SVKRNQTREF DGTEV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
IFT81 Polyclonal antibody specifically detects IFT81 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
IFT81 | |
Polyclonal | |
Unconjugated | |
IFT81 | |
AW060663; carnitine deficiency-associated gene expressed in ventricle 1; carnitine deficiency-associated protein expressed in ventricle 1; Cdv1; CDV-1; CDV1R; CDV-1R; DV1; Ift81; intraflagellar transport 81; intraflagellar transport 81 homolog; intraflagellar transport protein 81 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28981 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WYA0 | |
IFT81 | |
Recombinant protein corresponding to Human IFT81 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only