missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IFNW1 Recombinant Human Protein, hIgG1-Fc Tag

Expressed from DNA sequence encoding human IFNW1 (Cys 24-Ser 195) fused to Fc region of human IgG1 at N-terminus

Brand:  Sino Biological™ 10353H01H25

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13442979

  • £147.59 / Pack of 5

Estimated Shipment
02-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Measured in antiviral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV)
  • EC50 for effect is 0.1-0.5ng/mL
  • Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8 mM KH2
Specifications

Specifications

Performing cell based assays
46.6kd
5 x 5μg
Full Length, Recombinant
Interferon, omega-1
Assay Standard
Mammalian
hIgG1-Fc Tag
Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
IFNW1
Cytokines and Receptors
IFNW1 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.