missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IFNB1 Recombinant Mouse Protein, hIgG1-Fc Tag

Expressed from DNA sequence encoding extracellular domain of mouse IFNB1 (Met 1-Asn 182) fused to Fc region of human IgG1 at C-terminus

Brand:  Thermo Scientific™ 50708M02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15349196

  • £464.29 / 25µg


Estimated Shipment
07-08-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
15977
≥90% by SDS-PAGE
Mouse
<1 EU/μg
IFNB1
Other Proteins
IFNB1 Recombinant Mouse Protein
Lyophilized
46.7kd
5 x 5μg
Truncated, Recombinant
Interferon Beta 1, Fibroblast
Assay Standard
Mammalian
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.