missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IFNA4 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human IFNα 4a (Met 1-Asp 189) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10336H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15295205

  • £166.56 / 50µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
Specifications

Specifications

Analysis of protein structure and performing cell based assays
3441
≥85% by SDS-PAGE
Human
<1 EU/μg
IFNA4
Cytokines and Receptors
IFNA4 Recombinant Human Protein
Lyophilized
20.7kd
50μg
Full Length, Recombinant
Interferon Alpha 4
Assay Standard
Insect
His Tag
Videos