missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IFN-alpha 2 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ 131001
Code : PN
Additional Details : Weight : 0.10000kg
Description
Rat IFN alpha A Recombinant Protein, 131001, from Invitrogen., 131001, from Invitrogen. Species Reactivity: Human; Applications: ELISA, Immunoprecipitation, Western Blot.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis. Assay Used to Measure Bioactivity: Interferon was titrated with the use of the cytopathic effect inhibition assay as described [Rubinstein, S. et al. (1981), J. Virol. 3 7, 755-758]; [Familletti, P.C., et al (1981), Methods in Enzymology, Vol. 78, 387-394]. In this antiviral assay for interferon about 1 unit/mL of interferon is the quantity necessary to produce a cytopathic effect of 50%. As there is no international Rat IFN-alpha standard, units are determined with respect to the international reference standard for mouse interferon alpha/beta (mouse IFN-alpha/beta) provided by the National Institutes of Health. Each blood donation used for interferon production tested negative for: HBsAg, HIV I-II, HCV and VDRL. After receipt, this product should be kept at 4°C while lyophilized and at -70°C after reconstitution in sterile distilled water for retention of full activity. After reconstitution store at -70°C in aliquots. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Further dilute only in buffers containing protein such as 0.1% bovine serum albumin (BSA).Specifications
ELISA, Immunoprecipitation, Western Blot | |
298213 | |
5 | |
E. coli | |
RUO | |
Rat IFN alpha 2 | |
Rat | |
Rt | |
Recombinant | |
Recombinant |
Citric acid with 0.125M trehalose, NaCl and no preservative; pH 5 | |
Rat IFN alpha 2 | |
1x105units | |
4°C | |
Rat interferon Alpha, IFNA | |
Ifna2 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |