Learn More
IFN gamma Rat, V450, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB83-V450-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962558
  • £136.00 / 25µg
Estimated Shipment: 06-06-2024
to see stock.
Specifications
IFN gamma
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Ifng
Protein A/G Purified
RUO
15978
IgG1 κ
Flow Cytometry
XMG1.2
Violet Fluor 450 (V450)
NP_032363
Rat
25 μg
Primary
Store at 2-4°C.