missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma, Biotin, clone: XMG1.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-7311-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15370050

  • £62.18 / 50µg


Estimated Shipment date
from Supplier 27-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The XMG1.2 antibody reacts with mouse interferon (IFN) gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN gamma is a 20kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN gamma, in synergy with other cytokines such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.Applications Reported: XMG1.2 has been reported for use in ELISA and immunoblotting (WB).Applications Tested: The XMG1.2 antibody has been tested by ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IFN gamma
XMG1.2
Biotin
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
ELISA
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interferon-gamma, IFN-g, IFNg
IgG1
RUO
Primary
Murine
Videos