missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma Mouse anti-Human, PerCP-Cyanine5.5, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 45-7319-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599036

  • £97.25 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Specifications

Specifications

IFN gamma
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01579
Interferon gamma
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
4S.B3
PerCP-Cyanine5.5
IFNG
Liquid
IFNG
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3458
Videos
Documents

For Research Use Only.