missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN gamma Monoclonal Antibody (B27), Pacific Blue™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCIFG28
Code : 50
Additional Details : Weight : 0.00050kg
Description
IFN gamma Monoclonal antibody specifically detects IFN gamma in Human samples. It is validated for Flow Cytometry.Specifications
IFN gamma | |
Monoclonal | |
0.2 mg/mL | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P01579 | |
IFNG | |
Human IFN-gamma. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
B27 | |
Pacific Blue | |
IFNG | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
Mouse | |
Affinity chromatography | |
RUO | |
3458 | |
4° C | |
Liquid |