missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma, Alexa Fluor 488, clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-7319-41

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13537660

  • £70.41 / Pack of 25


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.Applications Reported: This 4S.B3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This 4S.B3 antibody has been pre-titrated and tested by flow cytometric analysis of activated human peripheral blood mononuclear cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IFN gamma
4S.B3
Alexa Fluor 488
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interferon-gamma
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.