missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma Armenian Hamster anti-Mouse, Functional Grade, Clone: H22, eBioscience™

Armenian Hamster Monoclonal Antibody

£59.00 - £184.00

Specifications

Antigen IFN gamma
Clone H22
Target Species Mouse
Conjugate Functional Grade
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15597816
View Documents
Affymetrix eBioscience
16-7411-81
50 μg £59.00
50µg View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
15507826
View Documents
Affymetrix eBioscience
16-7411-85
500 μg £184.00
500µg View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Specifications

Specifications

IFN gamma
Mouse
IgG
Monoclonal
Liquid
Antibody
4° C
H22
Functional Grade
Functional Assay, Neutralization
Primary
Monoclonal
PBS with no preservative; pH 7.2
RUO

For Research Use Only.