missing translation for 'onlineSavingsMsg'
Learn More

IFNAR1, PE, clone: MAR1-5A3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5945-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15567056

  • £74.36 / 25µg


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities.Applications Reported: This MAR1-5A3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MAR1-5A3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IFN alpha receptor 1 (IFNAR1)
MAR1-5A3
PE
Conjugated
IgG1
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos