Learn More
IFNAR1 Monoclonal Antibody (MAR1-5A3), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5945-38
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities. Applications Reported: The MAR1-5A3 antibody has been reported for use in neutralization of IFNAR1 bioactivity. Applications Tested: The Functional Grade Purified MAR1-5A3 antibody has been tested by LAL assay to verify low endotoxin levels, by bioassay for neutralization of mouse IFNAR-1bioactivity. The MAR1-5A3 antibody at 0.04 μg/mL has been found to inhibit by 50% the biological effects of 0.5 ng/mL mouse IFN-a2 (ND50), in an EMCV assay of L929 cell protection. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in Best Protocols® section. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).Specifications
IFNAR1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P33896 | |
Ifnar1 | |
Affinity chromatography | |
RUO | |
15975 | |
4° C | |
Liquid |
Functional Assay, Neutralization | |
MAR1-5A3 | |
Functional Grade | |
Ifnar1 | |
alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1 | |
Mouse | |
5 mg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.