missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil hemitartrate, Tocris Bioscience™

Non-competitive NMDA antagonist. Also σ ligand

Brand:  Tocris (Bio-Techne) 0545/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13487703

  • £93.15 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

NMDA receptor antagonist, acting at the polyamine site. Also an α-adrenergic vasodilator. σ2 ligand displaying about 3-fold selectivity over σ1 sites.
Specifications

Chemical Identifiers

23210-56-2
325.452
(1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
C21H27NO2
UYNVMODNBIQBMV-UHFFFAOYSA-N
3689

Specifications

Ifenprodil hemitartrate
>99%
(1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3
325.452
400.49
23210-56-2
C21H27NO2
UYNVMODNBIQBMV-UHFFFAOYSA-N
4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
3689
10mg
Videos