missing translation for 'onlineSavingsMsg'
Learn More

IEM 1754 dihydrobromide, Tocris Bioscience™

Voltage-dependent, open-channel blocker of AMPA currents

Brand:  Tocris (Bio-Techne) 419910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13444334

  • £124.20 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Voltage-dependent open-channel blocker of AMPA receptors. Selective between subtypes; blocks GluA2 subunit-lacking (Ca2+-permeable) receptors more potently than GluA2-containing receptors. Also potently blocks NMDA receptor channels. Less sensitive than IEM 1460 (Cat. No. 1460) at a holding potential between -40 and -80mV.
Specifications

Chemical Identifiers

162831-31-4
412.254
iem 1754 dihydrobromide, iem1754 dihydrobromide, iem 1754 2hbr, iem1754 2 hbr, 1-amino-5-1-adamantylmethylamino-pentane dihydrobromide, n1-adamantan-1-ylmethyl pentane-1,5-diamine dihydrobromide, adamantan-1-ylmethyl 5-aminopentyl amine dihydrobromide, n-tricyclo 3.3.1.13,7 dec-1-ylmethyl-1,5-pentanediamine dihydrobromide, 5-aminopentyl 3r,5s,7s-adamantan-1-ylmethyl amine dihydrobromide
N'-(1-adamantylmethyl)pentane-1,5-diamine;dihydrobromide
C16H32Br2N2
JMPHTNNJCZDROM-UHFFFAOYSA-N
9887867
C1C2CC3CC1CC(C2)(C3)CNCCCCCN.Br.Br

Specifications

IEM 1754 dihydrobromide
162831-31-4
iem 1754 dihydrobromide, iem1754 dihydrobromide, iem 1754 2hbr, iem1754 2 hbr, 1-amino-5-1-adamantylmethylamino-pentane dihydrobromide, n1-adamantan-1-ylmethyl pentane-1,5-diamine dihydrobromide, adamantan-1-ylmethyl 5-aminopentyl amine dihydrobromide, n-tricyclo 3.3.1.13,7 dec-1-ylmethyl-1,5-pentanediamine dihydrobromide, 5-aminopentyl 3r,5s,7s-adamantan-1-ylmethyl amine dihydrobromide
C1C2CC3CC1CC(C2)(C3)CNCCCCCN.Br.Br
412.254
412.25
10mg
C16H32Br2N2
JMPHTNNJCZDROM-UHFFFAOYSA-N
N'-(1-adamantylmethyl)pentane-1,5-diamine;dihydrobromide
9887867
>98%
Videos