Learn More
Tocris Bioscience™ IDRA 21
Inhibits AMPA receptor desensitization
Brand:  Tocris Bioscience™ 121910
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13448913
  • £169.05 / 10mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).
Specifications
Specifications
IDRA 21
>98
10mg
IDRA 21
232.68