Learn More
Tocris Bioscience™ IDRA 21
Inhibits AMPA receptor desensitization
Brand:  Tocris Bioscience™ 121950
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13438913
  • £557.72 / 50mg
Estimated Shipment
01-07-2019
Description
Description
Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).
Specifications
Specifications
IDRA 21
>98
50mg
IDRA 21
232.68