Learn More
IDO Recombinant Polyclonal Antibody, Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 711778
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Pig, Horse, Goat Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IDO Recombinant Polyclonal antibody specifically detects IDO in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
IDO | |
Recombinant Polyclonal | |
Unconjugated | |
IDO1 | |
EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
Rabbit | |
Protein A | |
RUO | |
3620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P14902 | |
IDO1 | |
Protein corresponding to human IDO1 (aa1-aa403) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |