Learn More
IDE 2, Tocris Bioscience™
Induces definitive endoderm formation in mouse and human ESCs
POA
Description
Description
Cell-permeable inducer of definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 223 nM for induction of Sox17 expression in ESCs). Reported to activate TGF-β signaling and downstream Smad2 phosphorylation; upregulates Nodal expression. Promotes pancreatic progenitor cell formation in vitro, and gut tube formation in vivo.