Learn More
IDE 2, Tocris Bioscience™
Induces definitive endoderm formation in mouse and human ESCs
Brand:  RD Systems (Bio-Techne) 4016/50
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 50mg
 View more versions of this product
Product Code. 13462794
  • £730.59 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Cell-permeable inducer of definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 223 nM for induction of Sox17 expression in ESCs). Reported to activate TGF-β signaling and downstream Smad2 phosphorylation; upregulates Nodal expression. Promotes pancreatic progenitor cell formation in vitro, and gut tube formation in vivo.
Specifications
Chemical Identifiers
1136466-93-7
240.303
ide 2, ide-2, heptanedioic acid-1-2-cyclopentylidenehydrazide, 7-2-cyclopentylidenehydrazino-7-oxoheptanoic acid, 7-oxo-7-2-cyclopentylidenehydrazino heptanoic acid, 6-n'-cyclopentylidenehydrazinecarbonyl hexanoic acid
7-(2-cyclopentylidenehydrazinyl)-7-oxoheptanoic acid
C12H20N2O3
CVYPYYPTFNVREL-UHFFFAOYSA-N
23836158
C1CCC(=NNC(=O)CCCCCC(=O)O)C1
Specifications
50mg
1136466-93-7
ide 2, ide-2, heptanedioic acid-1-2-cyclopentylidenehydrazide, 7-2-cyclopentylidenehydrazino-7-oxoheptanoic acid, 7-oxo-7-2-cyclopentylidenehydrazino heptanoic acid, 6-n'-cyclopentylidenehydrazinecarbonyl hexanoic acid
C1CCC(=NNC(=O)CCCCCC(=O)O)C1
240.303
240.3
IDE 2
C12H20N2O3
CVYPYYPTFNVREL-UHFFFAOYSA-N
7-(2-cyclopentylidenehydrazinyl)-7-oxoheptanoic acid
23836158
>98%