Learn More
IDE 1, Tocris Bioscience™
Induces definitive endoderm formation in mouse and human ESCs
Brand:  Tocris (Bio-Techne) 401550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13432794
  • £895.23 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Induces definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 125 nM). Thought to activate the TGF-β signaling pathway; induces Smad2 phosphorylation and increases levels of Nodal expression.
This compound is mixture of E/Z isomers.
Specifications
Chemical Identifiers
1160927-48-9
306.318
ide 1
2-[(6-carboxyhexanoylhydrazinylidene)methyl]benzoic acid
C15H18N2O5
ABKJCDILEUEJSH-UHFFFAOYSA-N
66974738
C1=CC=C(C(=C1)C=NNC(=O)CCCCCC(=O)O)C(=O)O
Specifications
50mg
1160927-48-9
ide 1
C1=CC=C(C(=C1)C=NNC(=O)CCCCCC(=O)O)C(=O)O
306.318
306.31
IDE 1
C15H18N2O5
ABKJCDILEUEJSH-UHFFFAOYSA-N
2-[(6-carboxyhexanoylhydrazinylidene)methyl]benzoic acid
66974738
>98%