missing translation for 'onlineSavingsMsg'
Learn More

Idazoxan hydrochloride, Tocris Bioscience™

α2 antagonist. Also I2 ligand

Brand:  Tocris (Bio-Techne) 079350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13468013

  • £401.31 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

α2-adrenoceptor antagonist, and I2 ligand, selective over I1 sites (pKi values are 5.90, 7.22, 8.01, 7.43, and 7.7 for I1, I2, α2A, α2B, and α2C receptors respectively).
Specifications

Chemical Identifiers

79944-56-2
240.687
idazoxan hydrochloride, idazoxan hcl, opera_id_847, d01znh, +/--idazoxan monohydrochloride, 1h-imidazole, 2-2,3-dihydro-1,4-benzodioxin-2-yl-4,5-dihydro-, monohydrochloride, 1h-imidazole, 2-2,3-dihydro-1,4-benzodioxin-2-yl-4,5-dihydro-,monohydrochloride, hcl ,, 2-1,4-benzodioxan-2-yl-2-imidazolinehydrochloride, +/--2-1,4-benzodioxan-2-yl-2-imidazoline hydrochloride
2-(2,3-dihydro-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
C11H13ClN2O2
MYUBYOVCLMEAOH-UHFFFAOYSA-N
154494
C1CN=C(N1)C2COC3=CC=CC=C3O2.Cl

Specifications

50mg
79944-56-2
idazoxan hydrochloride, idazoxan hcl, opera_id_847, d01znh, +/--idazoxan monohydrochloride, 1h-imidazole, 2-2,3-dihydro-1,4-benzodioxin-2-yl-4,5-dihydro-, monohydrochloride, 1h-imidazole, 2-2,3-dihydro-1,4-benzodioxin-2-yl-4,5-dihydro-,monohydrochloride, hcl ,, 2-1,4-benzodioxan-2-yl-2-imidazolinehydrochloride, +/--2-1,4-benzodioxan-2-yl-2-imidazoline hydrochloride
C1CN=C(N1)C2COC3=CC=CC=C3O2.Cl
240.687
240.69
Idazoxan hydrochloride
C11H13ClN2O2
MYUBYOVCLMEAOH-UHFFFAOYSA-N
2-(2,3-dihydro-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
154494
>98%
Videos
Documents