Learn More
ID2 Mouse anti-Human, Mouse, APC, Clone: ILCID2, eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen 17-9475-80
Additional Details : Weight : 0.01000kg
Description
Description: This ILCID2 monoclonal antibody recognizes mouse and human ID2. This ILCID2 antibody requires Foxp3/Transcription Factor Staining Buffer set and will perform poorly when used with standard IC Fixation & Permeabilization Buffer Set and protocol. Applications Reported: This ILCID2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ILCID2 antibody has been tested by flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.Specifications
ID2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P41136, Q02363 | |
Ac2-300; AI255428; BHLHB26; C78922; GIG8; Id-2; ID2; ID2A; ID2H; Idb2; inhibitor of DNA binding 2 | |
Mouse | |
IgG1, kappa | |
25 μg | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
15902, 3398 |
Flow Cytometry | |
ILCID2 | |
APC | |
ID2 | |
Liquid | |
Id2 | |
Purified recombit fragment of human ID2, corresponding to amino acids 1-134 expressed in E. coli | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human, Mouse |