Learn More
Alfa Aesar™ ICP-MS Soil Spike Standard solution for 6020 CLP-M, Specpure™
Brand: Alfa Aesar™ 042604.AE
Code : NEW
Additional Details : Weight : 0.00010kg Transport : UN number : 3264 Chem class : 1a Pack group : III
Specifications
ICP-MS Soil Spike Standard solution for 6020 CLP-M | |
Liquid | |
UN3264 | |
Matrix: 2% HNO3/tr. tartaric acid |
Specpure™ | |
MFCD02266211 | |
100mL |