missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ ICP-MS Internal Standard solution, Specpure™

Brand:  Alfa Aesar™ 044651.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

Product Code. 15452027

  • POA / 100mL

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

ICP-MS Internal Standard solution
Liquid
UN3264
Matrix: 5% HNO3
Specpure™
MFCD00803321
100mL
Videos
SDS