missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ICOS Recombinant Mouse Protein, hIgG1-Fc Tag

Expressed from DNA sequence encoding mouse ICOS extracellular domain (Met 1-Leu 142) fused to Fc region of human IgG1 at C-terminus

Brand:  Sino Biological™ 50466M02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15373276

  • £161.36 / 50µg


Estimated Shipment
15-05-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
54167
≥90% by SDS-PAGE
Mouse
<1 EU/μg
ICOS
CD Proteins
ICOS Recombinant Mouse Protein
Lyophilized
40.9kd
50μg
Truncated, Recombinant
Inducible T Cell Co-Stimulator
Assay Standard
Mammalian
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.