missing translation for 'onlineSavingsMsg'
Learn More

ICI 63197, Tocris Bioscience™

PDE4 inhibitor

Brand:  Tocris (Bio-Techne) 181650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460064

  • £311.64 / 50mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent phosphodiesterase (PDE) 4 inhibitor (IC50 = 35 nM for inhibition of [3H]-rolipram binding to rat brain). Antidepressant following systemic administration in vivo.
Specifications

Chemical Identifiers

27277-00-5
207.237
2-amino-6-methyl-4-propyl-1,2,4 triazolo 1,5-a pyrimidin-5 4h-one, caswell no. 033e, 2-amino-6-methyl-4-propyl-1,2,4 triazolo 1,5-a pyrimidin-5-one, unii-015g4i43yd, 2-amino-4,5-dihydro-6-methyl-4-propyl-s-triazolo-1,5-a-pyrimidin-5-one, 2-amino-6-methyl-5-oxo-4n-propyl-4,5-dihydro-s-triazolo 1,5-a pyrimidine, 1,2,4 triazolo 1,5-a pyrimidin-5 4h-one, 2-amino-6-methyl-4-propyl, 2-amino-4,5-dihydro-6-methyl-4-propyl-5-triazolo-1,5-c-pyramidin-5-one, 2-amino-6-methyl-4-propyl-1,2,4-triazolo 1,5-a pyrimidin-5 4h-one
2-amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5-one
C9H13N5O
UQDVRVNMIJAGRK-UHFFFAOYSA-N
62824
CCCN1C(=O)C(=CN2C1=NC(=N2)N)C

Specifications

50mg
27277-00-5
2-amino-6-methyl-4-propyl-1,2,4 triazolo 1,5-a pyrimidin-5 4h-one, caswell no. 033e, 2-amino-6-methyl-4-propyl-1,2,4 triazolo 1,5-a pyrimidin-5-one, unii-015g4i43yd, 2-amino-4,5-dihydro-6-methyl-4-propyl-s-triazolo-1,5-a-pyrimidin-5-one, 2-amino-6-methyl-5-oxo-4n-propyl-4,5-dihydro-s-triazolo 1,5-a pyrimidine, 1,2,4 triazolo 1,5-a pyrimidin-5 4h-one, 2-amino-6-methyl-4-propyl, 2-amino-4,5-dihydro-6-methyl-4-propyl-5-triazolo-1,5-c-pyramidin-5-one, 2-amino-6-methyl-4-propyl-1,2,4-triazolo 1,5-a pyrimidin-5 4h-one
CCCN1C(=O)C(=CN2C1=NC(=N2)N)C
207.237
207.23
ICI 63197
C9H13N5O
UQDVRVNMIJAGRK-UHFFFAOYSA-N
2-amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5-one
62824
>99%
Videos
Documents