missing translation for 'onlineSavingsMsg'
Learn More

ICI 199,441 hydrochloride, Tocris Bioscience™

Potent κ agonist

Brand:  Tocris (Bio-Techne) 077850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437993

  • £349.81 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent κ agonist, 146-fold more active than U-50488 (Cat. No. 0495) in vitro.
Specifications

Chemical Identifiers

115199-84-3
427.794
ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride
C21H25Cl3N2O
VFLWVWZSDBTGQJ-VEIFNGETSA-N
3082717
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

ICI 199, 441 hydrochloride
115199-84-3
ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl
427.794
427.8
50mg
C21H25Cl3N2O
VFLWVWZSDBTGQJ-VEIFNGETSA-N
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride
3082717
>99%
Videos