missing translation for 'onlineSavingsMsg'
Learn More

ICI 192,605, Tocris Bioscience™

Potent thromboxane A2/TP receptor antagonist

Brand:  Tocris (Bio-Techne) 083710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438063

  • £149.94 / 10mg

Estimated Shipment date
from Supplier 22-01-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent thromboxane A2 receptor (TP receptor) antagonist. Competitively inhibits contractile responses to U 46619 (Cat. No. 1932).
Specifications

Chemical Identifiers

117621-64-4
402.871
d0b9na, 6-2beta-2-chlorophenyl-6beta-2-hydroxyphenyl-1,3-dioxan-5beta-yl-4-hexenoic acid, e-6-2s,4s,5r-2-2-chlorophenyl-4-2-hydroxyphenyl-1,3-dioxan-5-yl hex-4-enoic acid
(E)-6-[(2S,4S,5R)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]hex-4-enoic acid
C22H23ClO5
WHUIENZXNGAHQI-BNGKFCBFSA-N
23654676
C1C(C(OC(O1)C2=CC=CC=C2Cl)C3=CC=CC=C3O)CC=CCCC(=O)O

Specifications

10mg
117621-64-4
d0b9na, 6-2beta-2-chlorophenyl-6beta-2-hydroxyphenyl-1,3-dioxan-5beta-yl-4-hexenoic acid, e-6-2s,4s,5r-2-2-chlorophenyl-4-2-hydroxyphenyl-1,3-dioxan-5-yl hex-4-enoic acid
C1C(C(OC(O1)C2=CC=CC=C2Cl)C3=CC=CC=C3O)CC=CCCC(=O)O
402.871
402.87
ICI 192, 605
C22H23ClO5
WHUIENZXNGAHQI-BNGKFCBFSA-N
(E)-6-[(2S,4S,5R)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]hex-4-enoic acid
23654676
>98%
Videos
Documents